COMH4211BA16 Combi-Blocks, Inc.

1-(2-Cyclopropylmethoxy-5-methyl-phenyl)-ethylamine

  • MDL number MFCD12138640
  • Cas Number : 1156176-99-6
  • Molecular Weight : 205.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6058

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6058
Availability details for
Quantity
  • Availability
  • Add to Cart