COMH4211BA17 Combi-Blocks, Inc.

5-Bromo-4-fluoro-2,n-dimethyl-benzamide

  • MDL number MFCD31579364
  • Cas Number : 2270911-42-5
  • Molecular Weight : 246.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6061

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6061
Availability details for
Quantity
  • Availability
  • Add to Cart