COMH4211BA19 Combi-Blocks, Inc.

5-Fluoro-2-(2-hydroxyethyl)benzonitrile

  • MDL number MFCD31579473
  • Cas Number : 2050016-94-7
  • Molecular Weight : 165.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6066

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6066
Availability details for
Quantity
  • Availability
  • Add to Cart