COMH4211BA21 Combi-Blocks, Inc.

3-Bromo-4-nitrobenzamide

  • MDL number MFCD18392262
  • Cas Number : 1261794-86-8
  • Molecular Weight : 245.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6084

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6084
Availability details for
Quantity
  • Availability
  • Add to Cart