COMH4211BA23 Combi-Blocks, Inc.

(4-Hydroxy-pyrazol-1-yl)-acetic acid benzyl ester

  • MDL number MFCD27931423
  • Cas Number : 884344-73-4
  • Molecular Weight : 232.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6093

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6093
Availability details for
Quantity
  • Availability
  • Add to Cart