COMH4211BA25 Combi-Blocks, Inc.

(2-(Benzyloxy)-5-iodophenyl)methanol

  • MDL number MFCD30578770
  • Cas Number : 636594-69-9
  • Molecular Weight : 340.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6097

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6097
Availability details for
Quantity
  • Availability
  • Add to Cart