COMH4211BA26 Combi-Blocks, Inc.

Trans 4-amino-cyclohexanecarboxylic acid ethylamide

  • MDL number MFCD31557888
  • Cas Number : 412290-83-6
  • Molecular Weight : 170.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6099

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6099
Availability details for
Quantity
  • Availability
  • Add to Cart