COMH4211BA29 Combi-Blocks, Inc.

1-(3-Bromo-2-methylphenyl)piperidin-2-one

  • MDL number MFCD31583068
  • Cas Number : 1231891-49-8
  • Molecular Weight : 268.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6108

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6108
Availability details for
Quantity
  • Availability
  • Add to Cart