COMH4211BA2A Combi-Blocks, Inc.

4-(Propane-2-sulfonylmethyl)-benzoic acid

  • MDL number MFCD12801115
  • Cas Number : 1183385-39-8
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6111

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6111
Availability details for
Quantity
  • Availability
  • Add to Cart