COMH4211BA2B Combi-Blocks, Inc.

Methyl 4-bromo-2-isopropoxybenzoate

  • MDL number MFCD25954231
  • Cas Number : 1016313-23-7
  • Molecular Weight : 273.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6113

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6113
Availability details for
Quantity
  • Availability
  • Add to Cart