COMH4211BA2C Combi-Blocks, Inc.

4-(4-Carboxymethoxy-2-fluoro-phenyl)-piperidine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD31583398
  • Cas Number : 2270906-29-9
  • Molecular Weight : 353.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6114

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6114
Availability details for
Quantity
  • Availability
  • Add to Cart