COMH4211BA2D Combi-Blocks, Inc.

3-(5-Chloro-2-methylphenyl)-3-oxopropanenitrile

  • MDL number MFCD24322975
  • Cas Number : 1205531-82-3
  • Molecular Weight : 193.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6115

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6115
Availability details for
Quantity
  • Availability
  • Add to Cart