COMH4211BA2E Combi-Blocks, Inc.

tert-Butyl 2-(4-bromophenylthio)-2-methylpropanoate

  • MDL number MFCD09863883
  • Cas Number : 247923-30-4
  • Molecular Weight : 331.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6117

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6117
Availability details for
Quantity
  • Availability
  • Add to Cart