COMH4211BA2F Combi-Blocks, Inc.

[(4-Fluoro-2-methylphenyl)methyl](propan-2-yl)amine

  • MDL number MFCD16686582
  • Cas Number : 1247737-67-2
  • Molecular Weight : 181.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6122

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6122
Availability details for
Quantity
  • Availability
  • Add to Cart