COMH4211BA32 Combi-Blocks, Inc.

3-(2,3,6-Trifluoro-phenyl)-propan-1-ol

  • MDL number MFCD09028667
  • Cas Number : 1057671-37-0
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6134

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6134
Availability details for
Quantity
  • Availability
  • Add to Cart