COMH4211BA35 Combi-Blocks, Inc.

3-(2,4,6-Trifluoro-phenyl)-propionaldehyde

  • MDL number MFCD09028566
  • Cas Number : 377084-11-2
  • Molecular Weight : 188.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6139

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6139
Availability details for
Quantity
  • Availability
  • Add to Cart