COMH4211BA37 Combi-Blocks, Inc.

tert-Butyl 4-allylpiperidine-1-carboxylate

  • MDL number MFCD09910235
  • Cas Number : 206446-47-1
  • Molecular Weight : 225.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6142

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6142
Availability details for
Quantity
  • Availability
  • Add to Cart