COMH4211BA38 Combi-Blocks, Inc.

Carbamic acid, (4-pentenyloxy)-, 1,1-dimethylethyl ester

  • MDL number MFCD24466958
  • Cas Number : 635757-94-7
  • Molecular Weight : 201.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6144

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6144
Availability details for
Quantity
  • Availability
  • Add to Cart