COMH4211BA39 Combi-Blocks, Inc.

3-(4-Fluorophenoxy)azetidine

  • MDL number MFCD09861914
  • Cas Number : 702628-84-0
  • Molecular Weight : 167.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6148

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6148
Availability details for
Quantity
  • Availability
  • Add to Cart