COMH4211BA3C Combi-Blocks, Inc.

Pellitorine

  • MDL number MFCD01735995
  • Cas Number : 18836-52-7
  • Molecular Weight : 223.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6158

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6158
Availability details for
Quantity
  • Availability
  • Add to Cart