COMH4211BA3E Combi-Blocks, Inc.

5-Bromo-3-chloro-2-(cyclopropylmethoxy)pyridine

  • MDL number MFCD28681177
  • Cas Number : 1431532-94-3
  • Molecular Weight : 262.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6161

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6161
Availability details for
Quantity
  • Availability
  • Add to Cart