COMH4211BA42 Combi-Blocks, Inc.

Prop-2-en-1-yl n-(6-aminohexyl)carbamate

  • MDL number MFCD24444612
  • Cas Number : 184292-16-8
  • Molecular Weight : 200.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6173

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6173
Availability details for
Quantity
  • Availability
  • Add to Cart