COMH4211BA43 Combi-Blocks, Inc.

2-Chloro-6-(pyrrolidine-1-carbonyl)pyridine

  • MDL number MFCD16698129
  • Cas Number : 720693-07-2
  • Molecular Weight : 210.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6174

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6174
Availability details for
Quantity
  • Availability
  • Add to Cart