COMH4211BA44 Combi-Blocks, Inc.

Ethyl 3-(2,5-dichloropyridin-4-yl)-3-oxopropionoate

  • MDL number MFCD31628840
  • Cas Number : 1020636-10-5
  • Molecular Weight : 262.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6176

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6176
Availability details for
Quantity
  • Availability
  • Add to Cart