COMH4211BA48 Combi-Blocks, Inc.

(S)-1-Cyclopropylmethyl-3-methyl-piperazine dihydrochloride

  • MDL number MFCD31539547
  • Cas Number : 2203714-62-7
  • Molecular Weight : 227.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6188

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6188
Availability details for
Quantity
  • Availability
  • Add to Cart