COMH4211BA49 Combi-Blocks, Inc.

(5-Amino-2-methoxyphenyl)(pyrrolidin-1-yl)methanone

  • MDL number MFCD11695344
  • Cas Number : 1071292-71-1
  • Molecular Weight : 220.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6190

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6190
Availability details for
Quantity
  • Availability
  • Add to Cart