COMH4211BA4B Combi-Blocks, Inc.

(5-Bromo-2-methoxy-pyridin-3-yl)-pyrrolidin-1-yl-methanone

  • MDL number MFCD26129088
  • Cas Number : 1707583-16-1
  • Molecular Weight : 285.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6193

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6193
Availability details for
Quantity
  • Availability
  • Add to Cart