COMH4211BA4C Combi-Blocks, Inc.

1-tert-Butyl 2-chloromethyl (2s)-pyrrolidine-1,2-dicarboxylate

  • MDL number MFCD29483671
  • Cas Number : 34614-72-7
  • Molecular Weight : 263.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6194

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6194
Availability details for
Quantity
  • Availability
  • Add to Cart