COMH4211BA4D Combi-Blocks, Inc.

N-(2-Aminoethyl)-2-(4-fluorophenyl)acetamide

  • MDL number MFCD09740632
  • Cas Number : 73965-91-0
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6197

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6197
Availability details for
Quantity
  • Availability
  • Add to Cart