COMH4211BA4E Combi-Blocks, Inc.

Glycine, n-[5-(1,2-dithiolan-3-yl)-1-oxopentyl]-

  • MDL number MFCD30595895
  • Cas Number : 14631-30-2
  • Molecular Weight : 263.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6200

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6200
Availability details for
Quantity
  • Availability
  • Add to Cart