COMH4211BA50 Combi-Blocks, Inc.

3-Hydroxypropyl p-methoxybenzyl thioether

  • MDL number MFCD04039565
  • Cas Number : 202665-68-7
  • Molecular Weight : 212.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6203

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6203
Availability details for
Quantity
  • Availability
  • Add to Cart