COMH4211BA51 Combi-Blocks, Inc.

2-tert-Butylsulfanyl-3-iodo-pyridine

  • MDL number MFCD31539535
  • Cas Number : 2203194-88-9
  • Molecular Weight : 293.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6205

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6205
Availability details for
Quantity
  • Availability
  • Add to Cart