COMH4211BA52 Combi-Blocks, Inc.

[1-(Ethanesulfonyl)piperidin-4-yl]methanol

  • MDL number MFCD11611184
  • Cas Number : 1082811-96-8
  • Molecular Weight : 207.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6208

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6208
Availability details for
Quantity
  • Availability
  • Add to Cart