COMH4211BA53 Combi-Blocks, Inc.

1-(4-Chlorobenzyl)-1h-1,2,4-triazole-3-carboxylic acid

  • MDL number MFCD31579374
  • Cas Number : 1781909-34-9
  • Molecular Weight : 237.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6213

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6213
Availability details for
Quantity
  • Availability
  • Add to Cart