COMH4211BA54 Combi-Blocks, Inc.

4-Bromo-2-methyl-benzoic acid 2,5-dioxo-pyrrolidin-1-yl ester

  • MDL number MFCD31557871
  • Cas Number : 2203195-04-2
  • Molecular Weight : 312.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6215

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6215
Availability details for
Quantity
  • Availability
  • Add to Cart