COMH4211BA55 Combi-Blocks, Inc.

(E)-Ethyl 3-((1r,4r)-4-(tert-butoxycarbonylamino)-cyclohexyl)acrylate

  • MDL number MFCD24369573
  • Cas Number : 1359965-84-6
  • Molecular Weight : 297.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6217

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6217
Availability details for
Quantity
  • Availability
  • Add to Cart