COMH4211BA57 Combi-Blocks, Inc.

4-Aminobicyclo[2.2.2]octane-1-carboxylic acid

  • MDL number MFCD20640191
  • Cas Number : 13595-17-0
  • Molecular Weight : 169.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6219

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6219
Availability details for
Quantity
  • Availability
  • Add to Cart