COMH4211BA58 Combi-Blocks, Inc.

4-Amino-2-ethoxy-benzoic acid ethyl ester

  • MDL number MFCD24646203
  • Cas Number : 910572-97-3
  • Molecular Weight : 209.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6221

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6221
Availability details for
Quantity
  • Availability
  • Add to Cart