COMH4211BA5A Combi-Blocks, Inc.

3-Methoxynaphthalen-2-amine

  • MDL number MFCD17012370
  • Cas Number : 67291-63-8
  • Molecular Weight : 173.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6229

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6229
Availability details for
Quantity
  • Availability
  • Add to Cart