COMH4211BA5B Combi-Blocks, Inc.

Benzene, 1-bromo-2-fluoro-4-(2-methoxyethoxy)-

  • MDL number MFCD21106933
  • Cas Number : 929884-35-5
  • Molecular Weight : 249.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6230

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6230
Availability details for
Quantity
  • Availability
  • Add to Cart