COMH4211BA5C Combi-Blocks, Inc.

tert-Butyl 2,4,5-trifluorobenzoate

  • MDL number MFCD22393629
  • Cas Number : 182875-05-4
  • Molecular Weight : 232.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6231

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6231
Availability details for
Quantity
  • Availability
  • Add to Cart