COMH4211BA5E Combi-Blocks, Inc.

(2-(2-[2-(4-Sulfamoyl-benzoylamino)-ethoxy]-ethoxy)-ethyl)-carbamic acid tert-butyl ester

  • MDL number MFCD31579397
  • Cas Number : 769154-02-1
  • Molecular Weight : 431.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6234

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6234
Availability details for
Quantity
  • Availability
  • Add to Cart