COMH4211BA60 Combi-Blocks, Inc.

2-Bromo-6-mesitylpyridine

  • MDL number MFCD24221086
  • Cas Number : 868372-36-5
  • Molecular Weight : 276.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6237

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6237
Availability details for
Quantity
  • Availability
  • Add to Cart