COMH4211BA62 Combi-Blocks, Inc.

3-Bromo-n-(cyanomethyl)-2-methylbenzamide

  • MDL number MFCD21296815
  • Cas Number : 1491360-65-6
  • Molecular Weight : 253.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6241

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6241
Availability details for
Quantity
  • Availability
  • Add to Cart