COMH4211BA64 Combi-Blocks, Inc.

Toluene-4-sulfonic acid 2-(2-benzyloxycarbonylamino-ethoxy)-ethyl ester

  • MDL number MFCD29059964
  • Cas Number : 144208-62-8
  • Molecular Weight : 393.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6245

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6245
Availability details for
Quantity
  • Availability
  • Add to Cart