COMH4211BA65 Combi-Blocks, Inc.

2(1H)-Pyridinone, 1-(difluoromethyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-

  • MDL number MFCD30830208
  • Cas Number : 1352152-41-0
  • Molecular Weight : 271.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6250

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6250
Availability details for
Quantity
  • Availability
  • Add to Cart