COMH4211BA68 Combi-Blocks, Inc.

3-(4-Hydroxymethyl-phenoxy)-azetidine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD31618931
  • Cas Number : 2270908-33-1
  • Molecular Weight : 279.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 3-(4-(hydroxymethyl)phenoxy)azetidine-1-carboxylate
Inchi Code 1S/C15H21NO4/c1-15(2,3)20-14(18)16-8-13(9-16)19-12-6-4-11(10-17)5-7-12/h4-7,13,17H,8-10H2,1-3H3
Inchi Key FXJOXSPBCFHZOD-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6260

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6260
Availability details for
Quantity
  • Availability
  • Add to Cart