COMH4211BA69 Combi-Blocks, Inc.

Methyl 3-(3-ethoxy-3-oxopropanoyl)benzoate

  • MDL number MFCD28107471
  • Cas Number : 371249-81-9
  • Molecular Weight : 250.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6261

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6261
Availability details for
Quantity
  • Availability
  • Add to Cart