COMH4211BA6A Combi-Blocks, Inc.

1-[3'-(2-Amino-ethyl)-biphenyl-3-yl]-ethanol hydrochloride

  • MDL number MFCD31557813
  • Cas Number : 2206608-53-7
  • Molecular Weight : 277.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6262

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6262
Availability details for
Quantity
  • Availability
  • Add to Cart