COMH4211BA6C Combi-Blocks, Inc.

Methyl 5-amino-4-hydroxy-2-(trifluoromethyl)benzoate

  • MDL number MFCD28647753
  • Cas Number : 1163281-02-4
  • Molecular Weight : 235.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6265

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6265
Availability details for
Quantity
  • Availability
  • Add to Cart